Order Entry
Belgium
ContactUsLinkComponent
Lactococcus lactis subsp. lactis WDCM 00016 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Lactococcus lactis subsp. lactis WDCM 00016 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Catalog # VT000167-10EA
Supplier:  Merck
CAS Number:  
Lactococcus lactis subsp. lactis WDCM 00016 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Catalog # VT000167-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Lactococcus lactis WDCM 00016 Vitroids™, 50000 - 150000 CFU mean value range
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Derived from CECT® strains.

  • Origin Strain No. CECT® 185
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Sigma-Aldrich Co. LLC