Order Entry
Belgium
ContactUsLinkComponent
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Catalog # 83646.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Water, HiPerSolv CHROMANORM® for UHPLC-MS
Catalog # 83646.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Clear colorless liquid:
    complying
  • Color (APHA Pt-Co):
    max. 5
  • Residue after evaporation:
    max. 0.7 ppm
  • Total organic carbon:
    max. 10 ppb
  • Aluminium (Al):
    max. 5 ppb
  • Calcium (Ca):
    max. 5 ppb
  • Iron (Fe):
    max. 5 ppb
  • Potassium (K):
    max. 5 ppb
  • Magnesium (Mg):
    max. 10 ppb
  • Sodium (Na):
    max. 25 ppb
  • Chloride (Cl):
    max. 10 ppb
  • Nitrate (NO3):
    max. 10 ppb
  • Phosphate (PO4):
    max. 10 ppb
  • Fluorescence 254/450 nm (as Quinine):
    max. 0.5 ppb
  • Fluorescence 365/450 nm (as Quinine):
    max. 0.5 ppb
  • UHPLC/MS ESI/APCI+ (as Reserpine):
    max. 2 ppb
  • UHPLC/MS ESI/APCI- (as Reserpine):
    max. 10 ppb
  • PEG Signal-to-noise ratio:
    max. 50
  • UHPLC-UV Gradient (210 nm):
    max. 2.0 mAU
  • UHPLC-UV Gradient (254 nm):
    max. 0.5 mAU
  • UV Absorbance 210nm (vs air):
    max. 0.01 AU
  • UV Absorbance 220 nm (vs air):
    max. 0.01 AU
  • UV Absorbance 230 nm (vs air):
    max. 0.01 AU
  • UV Absorbance 240 nm (vs air):
    max. 0.01 AU
  • UV Absorbance 254 nm (vs air):
    max. 0.005 AU
  • UV Absorbance 260 nm (vs air):
    max. 0.005 AU
  • UV Absorbance 280 nm (vs air):
    max. 0.005 AU

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :Filtered through a 0.1 micron filter.
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332