Order Entry
Belgium
ContactUsLinkComponent
Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Flammable
Toxic
Health
Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Catalog # 85802.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Catalog # 85802.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.9%
  • Identity (IR):
    complying
  • Clear colorless liquid:
    complying
  • Color (APHA Pt-Co):
    max. 10
  • Water(KF):
    max. 0.01% w/w
  • Residue after evaporation:
    max. 0.7 ppm
  • Acidity:
    max. 0.001 meq/g
  • Alkalinity:
    max. 0.0002 meq/g
  • Aluminium (Al):
    max. 5 ppb
  • Calcium (Ca):
    max. 5 ppb
  • Iron (Fe):
    max. 5 ppb
  • Potassium (K):
    max. 5 ppb
  • Magnesium (Mg):
    max. 5 ppb
  • Sodium (Na):
    max. 25 ppb
  • Fluorescence 254/450 nm (as Quinine):
    max. 0.5 ppb
  • Fluorescence 365/450 nm (as Quinine):
    max. 0.5 ppb
  • UHPLC/MS ESI/APCI+ (as Reserpine):
    max. 2 ppb
  • UHPLC/MS ESI/APCI- (as Reserpine):
    max. 10 ppb
  • PEG Signal-to-noise ratio:
    max. 50
  • UHPLC-UV Gradient (230 nm):
    max. 2.0 mAU
  • UHPLC-UV Gradient (254 nm):
    max. 0.5 mAU
  • UV Absorbance 210nm:
    max. 0.50 AU
  • UV Absorbance 215 nm:
    max. 0.40 AU
  • UV Absorbance 220 nm:
    max. 0.20 AU
  • UV Absorbance 230 nm:
    max. 0.10 AU
  • UV Absorbance 254 nm:
    max. 0.01 AU
  • UV Absorbance 260 nm:
    max. 0.005 AU
  • UV Absorbance 280 nm:
    max. 0.005 AU

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
 :Filtered through a 0.1 micron filter.
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595