Orders LinkContactUsLinkComponent
Dumont #3c forceps
Dumont #3c forceps
#3c, Inox, 110 mm, Standard, Straight, 0,17×0,1 mm
Catalog # FSCI11231-20
Marketsource
Supplier:   FINE SCIENCE TOOLS
Dumont #3c forceps
Catalog # FSCI11231-20
Supplier:  FINE SCIENCE TOOLS
Marketsource
Dumont #3c forceps
Dumont #3c forceps
#3c, Inox, 110 mm, Standard, Straight, 0,17×0,1 mm
Price
32,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    #3c
  • Length:
    110 mm
  • Material:
    Inox
  • Tip style:
    Standard
  • Tip size:
    0,17×0,1 mm
  • Tip shape:
    Straight