Orders LinkContactUsLinkComponent
Dumont #6 forceps
Dumont #6 forceps
#6, Inox, 115 mm, Standard, Angled, 0,17×0,1 mm
Catalog # FSCI11260-20
Marketsource
Supplier:   FINE SCIENCE TOOLS
Dumont #6 forceps
Catalog # FSCI11260-20
Supplier:  FINE SCIENCE TOOLS
Marketsource
Dumont #6 forceps
Dumont #6 forceps
#6, Inox, 115 mm, Standard, Angled, 0,17×0,1 mm
Price
47,50 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Type:
    #6
  • Length:
    115 mm
  • Material:
    Inox
  • Tip style:
    Standard
  • Tip size:
    0,17×0,1 mm
  • Tip shape:
    Angled