Orders LinkContactUsLinkComponent
Moria Iris forceps
Moria Iris forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Moria Iris forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Moria Iris forceps
Moria Iris forceps
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Type
    Material
    Length
    Tip style
    Tip shape
    Tip size
    Tip width
    Availability
    Price
  • MC31
    Inox
    100 mm
    Serrated
    Curved
    0,5×0,4 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,50 €
    152,50 €
    1 * 1 ST
       152,50 €
    Specifications:
    Marketsource
    Marketsource
  • MC32
    Inox
    100 mm
    Serrated
    Straight
    0,5×0,3 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 147,00 €
    147,00 €
    1 * 1 Pce
       147,00 €
    Specifications:
    Marketsource
    Marketsource
  • MC31B
    Inox
    100 mm
    Smooth
    Curved
    0,5×,04 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
    Specifications:
    Marketsource
    Marketsource
  • MC32B
    Inox
    100 mm
    Smooth
    Straight
    0,5×0,3 mm
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 140,50 €
    140,50 €
    1 * 1 Pce
       140,50 €
    Specifications:
    Marketsource
    Marketsource
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.