Orders LinkContactUsLinkComponent
Potts-Smith forceps
Potts-Smith forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Potts-Smith forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Potts-Smith forceps
Potts-Smith forceps
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip shape
    Tip size
    Availability
    Price
  • Stainless steel
    180 mm
    Serrated
    Straight
    1,25×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 24,50 €
    24,50 €
    1 * 1 Pce
       24,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    180 mm
    Serrated
    Curved
    1,25×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 34,00 €
    34,00 €
    1 * 1 Pce
       34,00 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    180 mm
    1×2 teeth
    Straight
    1,25×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 33,50 €
    33,50 €
    1 * 1 Pce
       33,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    180 mm
    1×2 teeth
    Curved
    1,25×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 36,00 €
    36,00 €
    1 * 1 Pce
       36,00 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    210 mm
    Serrated
    Straight
    1,8×1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 32,00 €
    32,00 €
    1 * 1 Pce
       32,00 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    210 mm
    Serrated
    Curved
    1,8×1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 37,50 €
    37,50 €
    1 * 1 Pce
       37,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    250 mm
    Serrated
    Straight
    2×1,6 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 46,50 €
    46,50 €
    1 * 1 Pce
       46,50 €
    Specifications:
    Marketsource
    Marketsource