Orders LinkContactUsLinkComponent
Pierse fixation forceps
Pierse fixation forceps
Stainless steel, Debakey serrations, Straight, 1,75×1,5 mm, 1,5 mm
Catalog # FSCI18155-13
Marketsource
Supplier:   FINE SCIENCE TOOLS
Pierse fixation forceps
Catalog # FSCI18155-13
Supplier:  FINE SCIENCE TOOLS
Marketsource
Pierse fixation forceps
Pierse fixation forceps
Stainless steel, Debakey serrations, Straight, 1,75×1,5 mm, 1,5 mm
Price
146,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Material:
    Stainless steel
  • Tip style:
    Debakey serrations
  • Tip size:
    1,75×1,5 mm
  • Tip width:
    1,5 mm
  • Tip shape:
    Straight