Orders LinkContactUsLinkComponent
Plastic replacement tips
Plastic replacement tips
Marketsource
Supplier:   FINE SCIENCE TOOLS
Plastic replacement tips
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Plastic replacement tips
Plastic replacement tips
2 Options Available Below
Product Details & Documents
Pack of (2) PVDF tips and (3) screws. Tips are made of polyvinylidene fluoride (PVDF) and are electrostatic discharge (ESD) safe.
Compatible with Item No. 11980-13.
Product Family Options
Product Information
  • Material
    Length
    Tip size
    Availability
    Price
  • FSCI11988-03
    Plastic replacement tips for forceps, curved, very fine
    Plastic replacement tips for forceps, curved, very fine
    Stainless steel
    40 mm
    Ø 0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,50 €
    8,50 €
    1 * 1 ST
       8,50 €
    Specifications:
    Marketsource
    Marketsource
  • FSCI11988-02
    Plastic replacement tips for forceps, straight, very fine
    Plastic replacement tips for forceps, straight, very fine
    Stainless steel
    40 mm
    Ø 0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,50 €
    8,50 €
    1 * 1 ST
       8,50 €
    Specifications:
    Marketsource
    Marketsource