Orders LinkContactUsLinkComponent
Ring forceps
Ring forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Ring forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Ring forceps
3 Options Available Below
Product Details & Documents
For transport of tissue and organs. A limit screw prevents excessive applied pressure.
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip shape
    Tip size
    Availability
    Price
  • Stainless steel
    85 mm
    Smooth
    Straight
    1,5×0,45 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 66,50 €
    66,50 €
    1 * 1 Pce
       66,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    90 mm
    Smooth
    Straight
    3×0,75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,00 €
    54,00 €
    1 * 1 ST
       54,00 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    90 mm
    Smooth
    Straight
    6×0,9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,00 €
    57,00 €
    1 * 1 ST
       57,00 €
    Specifications:
    Marketsource
    Marketsource