Order Entry
Belgium
Orders LinkContactUsLinkComponent
Russian forceps

Russian forceps

Stainless steel, 150 mm, Cupped teeth, Straight, 6×11 mm
Catalog # FSCI11026-15
Marketsource
Supplier:   FINE SCIENCE TOOLS
Russian forceps
Catalog # FSCI11026-15
Supplier:  FINE SCIENCE TOOLS
Marketsource
Russian forceps
Russian forceps

Stainless steel, 150 mm, Cupped teeth, Straight, 6×11 mm

Price
52,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    150 mm
  • Material:
    Stainless steel
  • Tip style:
    Cupped teeth
  • Tip size:
    6×11 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.