Orders LinkContactUsLinkComponent
Taylor forceps
Taylor forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Taylor forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Taylor forceps
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip shape
    Tip size
    Availability
    Price
  • Stainless steel
    170 mm
    Serrated
    Curved
    1,5×1,05 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 40,50 €
    40,50 €
    1 * 1 Pce
       40,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    170 mm
    Serrated
    Straight
    1,5×1,05 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 35,50 €
    35,50 €
    1 * 1 Pce
       35,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    170 mm
    Smooth
    Curved
    1,5×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 36,50 €
    36,50 €
    1 * 1 Pce
       36,50 €
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    170 mm
    Smooth
    Straight
    1,5×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 30,50 €
    30,50 €
    1 * 1 Pce
       30,50 €
    Specifications:
    Marketsource
    Marketsource