Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bone Cutters and Rongeurs
Friedman rongeur
Friedman rongeur
FSCI16000-14
:
FINE SCIENCE TOOLS
Friedman rongeur
FSCI16000-14
:
FINE SCIENCE TOOLS
Type:
Curved
Description:
Friedman rongeurs, 2,5 mm cup diameter
Length:
130 mm
Pk:
1
These rongeurs are made of stainless steel.
Single joint action