Luer rongeur
Catalog # FSCI16001-15
Supplier: FINE SCIENCE TOOLS
Specifications
- Type:Curved
- Description:Luer rongeurs, 4 mm cup diameter
- Length:150 mm
- Pk:1
Specifications
About this item
These rongeurs are made of stainless steel.
- Single joint action