Order Entry
Belgium
ContactUsLinkComponent
Headspace oven and sample changer for coulometric KF titration
Headspace oven and sample changer for coulometric KF titration
Supplier:  SI Analytics
Certificates
undefined
Headspace oven and sample changer for coulometric KF titration
Supplier:  SI Analytics

Specifications

  • Catalog No:
  • SCOI285227850
  • SCOI285227840
  • SCOI285227820
  • Measuring range:
  • 1 μg - 100 mg absolute
  • 1 μg - 100 mg absolute
  • 1 μg - 100 mg absolute
  • Resolution:
  • 0,1 μg
  • 0,1 μg
  • 0,1 μg
  • Dosing volume:
  • Headspace vials (5 ml)
  • Headspace vials (5 ml)
  • Headspace vials (5 ml)
  • Temperature range:
  • 35...280 °C (isotherm)
  • 35...280 °C (isotherm)
  • 35...280 °C (isotherm)
  • W×D×H:
  • 300×240×450 mm
    420×460×450 mm with TO 7280
  • 300×240×450 mm
    420×460×450 mm with TO 7280
  • 300×240×450 mm
    420×460×450 mm with TO 7280
  • Weight:
  • 10 kg without/17 kg with TO 7280
  • 10 kg without/17 kg with TO 7280
  • 10 kg without/17 kg with TO 7280

Specifications

Frequently Bought Together