Order Entry
Belgium
Orders LinkContactUsLinkComponent
Laser microdissection (LMD) system

Laser microdissection (LMD) system

Laser microdissection system, LMD6, Diode pumped, solid state, 355 nm
Catalog # 630-3398
Supplier:   LEICA MICROSYSTEMS
Laser microdissection (LMD) system
Catalog # 630-3398
Supplier:  LEICA MICROSYSTEMS
Laser microdissection (LMD) system
Laser microdissection (LMD) system

Laser microdissection system, LMD6, Diode pumped, solid state, 355 nm

Price
238 652,35 €
Log in for availability
1 * 1 SET
Product Details & Documents

We move the laser, not the sample. And we use gravity for collection. LMD systems provides perfectly cut, contamination-free, analysis-ready dissectates.

  • Unique laser guidance based dissection method to collect dissectates via gravity – contact- and contamination-free
  • High precision optics to steer the laser beam along the desired cut line, leaving clean cuts of contamination-free isolates
  • LMD SmartCut objectives made specifically for the LMD system
  • Save on consumables, as the Leica LMD utilises standard PCR tubes to capture isolated samples - you will not be locked into expensive consumables with this system

Laser microdissection (LMD, also known as laser capture microdissection or LCM) enables users to isolate specific single cells or entire areas of tissue. Powered by a unique laser design and dynamic software, Leica LMD systems allow users to easily isolate regions of interest (ROI) from entire areas of tissue down to single cells or even subcellular structures such as chromosomes.

LMD is typically used in genomics (DNA), transcriptomics (mRNA, miRNA), proteomics, metabolomics, and even next generation sequencing (NGS). Researchers in neurology, cancer research, plant analysis, forensics or climate research rely on this method. Furthermore, LMD is a perfect tool for live cell culture (LCC), for cloning and re-cultivation, manipulation or downstream analysis.

Certifications: CE and CAS.

Ordering information: LMD slides are also available; contact your Avantor representative for more information. For more information on LMD and how we can help with sample preparation of downstream genomic and proteomic analyses, please contact your local Avantor representative.

Delivery information: Leica Laser Microdissection (LMD6) System Package includes DM6 upright microscope with motorised Z, CTR6 LED, 7-positon motorised nosepiece for objectives, mortised condenser, S28 condenser head (0,55 N.A., 28 mm working distance), motorised stage, Basic XYZ-module LX, Smart Move to control motorised stage, objectives (UVI 2,5×/0,07 N.A. Microdissection, UVI 5×/0,12 N.A. Microdissection, HC PL FLUOTAR 10×/0,32 N.A. and HCX PL FL L 20x/0,40 N.A. with correction collar), Patho diffusor plate for 1,25-5× object, Wacom Cintiq 22HD Pen Display, LMD CC7000 camera 11541544, C-mount HC 0,55×, Tube adapter including one documentation port. Purchase of LMD package includes installation and training.

Specifications
  • Model:
    LMD6
  • Wavelength:
    355 nm
  • Description:
    Laser microdissection system
  • Seq:
    02
  • Type:
    Diode pumped, solid state
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.