ContactUsLinkComponent
Safety helmet with CR2 reflective decals, EVOLite®
Safety helmet with CR2 reflective decals, EVOLite®
Catalog # JSPAAJB170-400-100
Supplier:  JSP
undefined
Safety helmet with CR2 reflective decals, EVOLite®
Catalog # JSPAAJB170-400-100
Supplier:  JSP
Specifications
  • Description:
    EVOLite® Safety helmet with CR2 reflective decals
  • Pk:
    1
Specifications
About this item
EVOLite® is one of the lightest safety helmets for sale in the UK, conforming to EN 397. This model is fitted with CR2 reflective decals.
  • Lightweight industrial safety helmet fitted with CR2 reflective decals
  • Tough ABS shell
  • Vented design
  • Wheel ratchet adjustment
  • Chamlon sweatband
The EVOLite® safety helmet is lightweight, extremely comfortable, and fits the widest range of head shapes and sizes. The harness and shell are truly integrated as one to give the best performance by keeping it firmly rooted on the head, even when fully inverted. Fitted with CR2 reflective decals which offer on average 60% more reflectivity than standard reflective clothing.
Weighing less than 300 g (dependent on model), the EVOLite® is the lightest safety helmet for sale in the UK. A 6-point terylene cradle harness system offers unrivalled comfort without compromising performance. Egyptian cotton core with porous PU coating for maximum sweat absorption. pH neutral, dermatologically tested. 3-D precision fitting uses the unique 1-2-3 point harness depth settings.
Certifications: EN 397 and EN 12899-1 (reflective).