Orders LinkContactUsLinkComponent
Junior IV goggle
Junior IV goggle
Junior IV goggle, Polycarbonate
Catalog # JSPAAGT020-141-300
Supplier:   JSP
Junior IV goggle
Catalog # JSPAAGT020-141-300
Supplier:  JSP
Junior IV goggle
Junior IV goggle
Junior IV goggle, Polycarbonate
Price
5,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Junior goggle with reduced size housing, fully conforming to EN 166 34B.
  • Junior goggle
  • Reduced size housing
  • One-piece polycarbonate lens
  • Elastic headband
The lens and body offer protection against medium energy, medium-speed particles at 120 m/s, large dust particles, and liquid droplets. The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Certifications: EN 166, EN 170.
Documents
Specifications
  • Description:
    Junior IV goggle
  • Lenses:
    Polycarbonate