Double lens goggle, EVO®
EVO® double lens goggle, PC outer lens, Acetate inner lens
Catalog # JSPAAGM020-823-000
Supplier: JSP
Jump to:
Product Details & Documents
Double lens goggle engineered to offer outstanding performance, compatibility, comfort, and looks.
- Double lens design
- Indirect venting
- PVC frame
- Elastic headband
- Contour design
- High level of protection
- Fully compatible with the JSP EVO® Range of industrial safety helmets
Double lens design for optimum anti fog protection, as well as anti-mist and scratch resistant lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Googles provide dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Certifications: EN 166; EN 170.
Documents
Manual Safety Eyewear
Manual
TDS Goggles Evo Double Lens
Datasheet
Specifications
- Description:EVO® double lens goggle
- Lenses:PC outer lens, Acetate inner lens
Accessories
Product Information
- Catalog #AvailabilityPrice
- RX prescription insert for EVO® goggles, TR 90 material, 6 g, 6 mm curveRX prescription insert for EVO® goggles, TR 90 material, 6 g, 6 mm curveJSPAAGU230-001-300Log in for availability1 * 1 ST - 5,80 €