ContactUsLinkComponent
Splash guard, without impact protection, Force®8
Splash guard, without impact protection, Force®8
Catalog # JSPAAFN060-000-000
Supplier:  JSP
undefined
Splash guard, without impact protection, Force®8
Catalog # JSPAAFN060-000-000
Supplier:  JSP
Specifications
  • Description:
    Force®8 splash guard
  • Pk:
    10
Specifications
About this item
The Force®8 Splash Guard can be fitted to a Force®8 half mask to reduce the likelihood of exhaled bacteria/virus hitting the wearer's face.
  • Easily fits to JSP Force®8 half mask
  • Provides a layer of protection to block aerosols and droplets
  • Prevents wearer from touching eyes and face
  • Quick and simple cleaning with soap and water
The Force®8 Splash Guard prevents the wearer from touching eyes and face. Quick and simple cleaning can be performed with soap and water.