ContactUsLinkComponent
Industrial browguards, EVOGuard® C
Industrial browguards, EVOGuard® C
Catalog # JSPAADB280-000-400
Supplier:  JSP
undefined
Industrial browguards, EVOGuard® C
Catalog # JSPAADB280-000-400
Supplier:  JSP
Specifications
  • Description:
    EVOGuard® C industrial browguard
  • Pk:
    1
Specifications
About this item
EVOGuard®C Industrial Browguard is designed for use with EVOGuard® visors, to provide innovative solutions to protect the wearer in a multitude of high-risk working environments.
  • Range of visors available
  • Compatible with other PPE products
  • Made of 30% glass filled nylon
  • JSP Revolution® wheel ratchet for a secure fit
  • 6-position harness
  • Recyclable
Available with a range of high performance visors compatible with EVOGuard® browguard for medium energy B-rated impact protection. The self-lubricating nylon hinges hold the visor securely in position even during extreme movement, whilst offering balanced, even weight distribution. This browguard features the new JSP Revolution® wheel ratchet, securely tightening and holding the unit firmly in position, no matter the task or amount of user movement involved. System comes complete with a wraparound, soft foam backed, replaceable, Chamlon sweatband which offers superior comfort.
The EVOGuard® range has been specifically designed to maximise compatibility with a range of other JSP products, such as the Force®8 respirator with PressToCheck™, Springfit™ disposable masks and the Sonis® helmet mounted ear defender range.
Certifications: EN 166, EN 1731.