Order Entry
Belgium
ContactUsLinkComponent
Recombinant human deiodinase 2
Recombinant human deiodinase 2
Supplier:  SELENOZYME
Certificates
undefined
Recombinant human deiodinase 2
Supplier:  SELENOZYME

Specifications

  • Storage conditions:
    Protein stored in Tris-EDTA buffer with 50% glycerol
  • Purity:
    ≥ 95%

Specifications

Related Information

Frequently Bought Together