Order Entry
Belgium
ContactUsLinkComponent
Recombinant Rat Thioredoxin Reductase 1
Recombinant Rat Thioredoxin Reductase 1
Supplier:  SELENOZYME
Certificates
undefined
Recombinant Rat Thioredoxin Reductase 1
Supplier:  SELENOZYME

Specifications

  • Storage conditions:
    Protein stored in Tris-EDTA buffer with 50% glycerol
  • Purity:
    ≥ 95%
  • Concentration:
    Concentration 1 mg protein/ml buffer.

Specifications

Related Information

Frequently Bought Together