ContactUsLinkComponent
Screw-in fittings PG, BOLA
  BOHLB151-03
 :  Bohlender
undefined
Screw-in fittings PG, BOLA
  BOHLB151-03
 :  Bohlender
  • Thread:
    1/2" NPT
  • Description:
    Screw-in fitting, wrench size 24 mm, PG thread 13,5
  • Pk:
    1
For connection of sensors with male thread PG to BOLA reactor lids.
  • For assembly on BOLA reactor lids
  • With hexagonal gripping surface in standard wrench size
  • Made of PTFE
For tight and safe insertion of sensors with male thread PG.