Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Microplate Seals
Closure for SuperFlex™ 96-well PCR plates
Closure for SuperFlex™ 96-well PCR plates
Closure for SuperFlex™ 96-well PCR plates
Catalog # SIMPT321-96N
Supplier:
Simport Scientific
Expand image
Closure for SuperFlex™ 96-well PCR plates
Catalog # SIMPT321-96N
Supplier:
Simport Scientific
Specifications
Colour:
Clear
Description:
Closure for SuperFlex™ 96-well PCR plates
Material:
PP
Pk:
100
Specifications
About this item
Product Details
Product Details