Orders LinkContactUsLinkComponent
Filtration kits, Sartoclear® Dynamics Lab V
Filtration kits, Sartoclear® Dynamics Lab V
   Sustainable Options Available
Fournisseur:   Sartorius
Filtration kits, Sartoclear® Dynamics Lab V
Fournisseur:  Sartorius
   Sustainable Options Available
Certificates
Filtration kits, Sartoclear® Dynamics Lab V
10 Options Available Below
Product Details & Documents
Sartoclear Dynamics® Lab V kits enable clarification and sterile filtration to be performed in a single step.
  • Clarify high-density mammalian cell culture in minutes
  • Out-of-the-box solution
  • Fast and effortless
  • No need for centrifugation
These kits simplify the process by fully eliminating the centrifugation step otherwise needed for clarification. As a result, they enable harvested cell cultures to be efficiently clarified and sterilised in minutes – quickly and easily.
Sartoclear Dynamics® Lab V kits are designed for simultaneous clarification and sterile filtration of ≻15 ml to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.
Specifications for Product Family
Specifications
  • Catalog No:
  • 514-1331
  • 514-1332
  • 514-1333
  • 514-1334
  • 514-1335
  • 514-1336
  • 514-1337
  • 514-1338
  • 514-1339
  • 514-1340
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Product Information
  • Membrane
    Capacity
    Pore size
    Disponibilité
    Prix
  • 514-1331
    Sartoclear® Dynamics Lab V, 2 g
    Sartoclear® Dynamics Lab V, 2 g
    Asymmetric PES
    150 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 206,93 €
    206,93 €
    1 * 12 ST
       206,93 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1331
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1332
    Sartoclear® Dynamics Lab V, 5 g
    Sartoclear® Dynamics Lab V, 5 g
    Asymmetric PES
    150 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 444,80 €
    444,80 €
    1 * 24 ST
       444,80 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1332
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1333
    Sartoclear® Dynamics Lab V, 5 g
    Sartoclear® Dynamics Lab V, 5 g
    Asymmetric PES
    250 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 476,80 €
    476,80 €
    1 * 24 ST
       476,80 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1333
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1334
    Sartoclear® Dynamics Lab V, 10 g
    Sartoclear® Dynamics Lab V, 10 g
    Asymmetric PES
    250 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 540,80 €
    540,80 €
    1 * 24 ST
       540,80 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1334
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1335
    Sartoclear® Dynamics Lab V, 5 g
    Sartoclear® Dynamics Lab V, 5 g
    Asymmetric PES
    500 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 604,80 €
    604,80 €
    1 * 24 ST
       604,80 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1335
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1336
    Sartoclear® Dynamics Lab V, 10 g
    Sartoclear® Dynamics Lab V, 10 g
    Asymmetric PES
    500 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 667,73 €
    667,73 €
    1 * 24 ST
       667,73 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1336
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1337
    Sartoclear® Dynamics Lab V, 20 g
    Sartoclear® Dynamics Lab V, 20 g
    Asymmetric PES
    500 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 378,67 €
    378,67 €
    1 * 12 ST
       378,67 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1337
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1338
    Sartoclear® Dynamics Lab V, 10 g
    Sartoclear® Dynamics Lab V, 10 g
    Asymmetric PES
    1000 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 955,73 €
    955,73 €
    1 * 24 ST
       955,73 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1338
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1339
    Sartoclear® Dynamics Lab V, 20 g
    Sartoclear® Dynamics Lab V, 20 g
    Asymmetric PES
    1000 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 502,40 €
    502,40 €
    1 * 12 ST
       502,40 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1339
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 514-1340
    Sartoclear® Dynamics Lab V, 40 g
    Sartoclear® Dynamics Lab V, 40 g
    Asymmetric PES
    1000 ml
    0,22 µm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 557,87 €
    557,87 €
    1 * 12 ST
       557,87 €
    Specifications:
      Low Manufacturing Impact
    Plus
    Specifications
    Cat. No.
    514-1340
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact