ContactUsLinkComponent
Chromogenic endotoxin quant kit, Pierce™
Chromogenic endotoxin quant kit, Pierce™
Certificates
Chromogenic endotoxin quant kit, Pierce™
Certificates
Specifications
  • Detection method:
    End-point Chromogenic Assay, Colorimetric
  • Range:
    0,01 - 0,1 EU/ml, 0,1 - 1,0 EU/ml
  • Sensitivity:
    0,01 EU/ml