Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Disc sensor, b.safe

Disc Sensor, with plug connection
Catalog # BOHLR640-05
Supplier:   Bohlender
Disc sensor, b.safe
Catalog # BOHLR640-05
Supplier:  Bohlender
Disc sensor, b.safe
Disc sensor, b.safe

Disc Sensor, with plug connection

Price
340,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Optional device for fill level or empty level control in containers and canisters made of non-conductive materials.

  • Avoid canister spills by connecting to b.safe Mini-Display or b.safe Display for electronic level control
  • Signal cables to connect the Disc Sensor with an alarm box are sold separately

The sensor is attached to the outside of a container with the included fastening material (adhesive hook-and-look strips or hook-and-look tape) at a designated level monitoring height. Via a signal cable, the sensor is connected with an alarm box (b.safe Mini-Display or b.safe Display). Not suitable for use in explosive areas or with an Isolated Amplifier.

 : Hook-and-loop tape or strips can be threaded into side brackets (tape) or simply stuck to container and sensor (strips). For emptying, the sensor can easily be removed and later reattached again to the correct position, or repositioned entirely.

 : The signal cables are not included in delivery, and must be ordered separately.

Specifications
  • Description:
    Disc Sensor, with plug connection
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Hook-and-loop tape, 10 mm × 2 m
    Hook-and-loop tape, 10 mm × 2 m
    BOHLR647-02
    Log in for availability
    1 * 1 ST - 33,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Hook-and-loop Strips, 25×25 mm
    Hook-and-loop Strips, 25×25 mm
    BOHLR648-01
    Log in for availability
    1 * 1 SET - 13,90 €