VWR® Sliding Tray Racks for Cryoboxes, Height 50 mm
Fournisseur: VWR Collection
Certificats
À propos de cet article
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
- Different height and depth dimensions are available and can be used in all freezer brands
- Racks for standard 2" cryoboxes (133×133×51 mm)
- Made from corrosion resistant Stainless steel with smoothed edges
These racks ensure an ergonomic operating process as the trays are placed in the rack. With a pull-out design it makes it easy to get samples without taking the whole rack out from the freezer. The tray has an easy access handle.
Measure the inside height and depth in the compartment of your freezer before choosing the right racks. In 'Related information' tab you will find additional information to support you in the selection.
Informations de commande: Delivered without cryoboxes.
Informations associées
Avantor Cell Culture Solutions.pdf
Documentation produit
Freezer racks – 2 boxes to optimize the storage capacity in VWR Eco Premium 264, 352 and 528
Détails supplémentaires sur le produit
Frequently Bought Together