Orders LinkContactUsLinkComponent
Elbow fitting, b.safe
Elbow fitting, b.safe
Elbow fitting, PE, GL 25 (male), GL 25 (female)
Catalog # BOHLR1224-01
Supplier:   Bohlender
Elbow fitting, b.safe
Catalog # BOHLR1224-01
Supplier:  Bohlender
Elbow fitting, b.safe
Elbow fitting, b.safe
Elbow fitting, PE, GL 25 (male), GL 25 (female)
Price
52,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Screw cap for thread GL 25 made of PPS with male thread GL 25.
  • Made of static dissipative PE-EX
  • With rotary elbow fitting
  • Without sagging
Angle of approx. 90° for a space-saving connection of b.safe Waste Tubes to b.safe Desktop Mounts or b.safe Drain Basin. Rotary elbow fitting for freely adapting the direction of the tube connection.
Specifications
  • Thread:
    GL 25 (male), GL 25 (female)
  • Description:
    Elbow fitting
  • Material:
    PE