T-connection tubes for waste pipes, b.safe
Catalog # BOHLR1192-01
Fournisseur: Bohlender
Spécifications
- Description:T-connection tubes for waste pipes, PE, with 3× clamp connection Ø 32 mm, total length 110 mm
- Pk:1
Spécifications
À propos de cet article
T-shaped connector made of static dissipative PE-EX.
With 3× union nut and clamp ring to insert and clamp b.safe Waste Pipes with Ext.Ø 32 mm.