Cryogenic warning labels
About this item
Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.
- Water resistant
- Wear and tear-proof
- Laminated
- No perforation
The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.
13 Options Available Below
Product Details & Documents
Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.
- Water resistant
- Wear and tear-proof
- Laminated
- No perforation
The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.
Product Family Options
Product Information
- MaterialLegendDimensionsPackedAvailabilityPrice
- GAINWL-012-1PBiohazard symbol labels
- GAINWL-013-1PRemovable warning symbol labels, blackRemovable warning symbol labels, blackPlasticRadioactive Picto12,7 mm1000 labels/rollLog in for availability1 * 1 ROU42,70 €
- GAINWL-008-1RRemovable warning symbol labelsRemovable warning symbol labelsPlasticHot Surface Hazard Picto19,05×19,05 mm250 labels/rollLog in for availability1 * 1 ROU178,00 €
- GAINWL-009-1RRemovable warning symbol labelsRemovable warning symbol labelsPlasticGeneral Warning Picto19,05×19,05 mm250 labels/rollLog in for availability1 * 1 ROU178,00 €
- GAINWL-010-1RRemovable warning symbol labelsRemovable warning symbol labelsPlasticLow Temperature Picto19,05×19,05 mm250 labels/rollLog in for availability1 * 1 ROU178,00 €
- GAINWL-016-0.5PHmis warning labels
- GAINWL-011-1PStop symbol for frozen surfaces labelsStop symbol for frozen surfaces labelsPlastic—38,1×38,1 mm1000 labels/rollLog in for availability1 * 1 ROU178,00 €
- GAINWL-001-0.25PToxic text labels
- GAINWL-002-0.25PNon-hazardous labels
- GAINWLA-007-1PBiohazard, yellow background labelsBiohazard, yellow background labelsPlasticBiohazard Picto44,45×19,05 mm250 labels/rollLog in for availability1 * 1 ROU137,00 €
- GAINWLA-006-0.1PBiohazard labelsBiohazard labelsPlasticBiohazard Picto50,8×50,8 mm100 labels/rollLog in for availability1 * 1 ROU45,60 €
- GAINWL-015-0.5PHmis warning labels
- GAINWL-004-1PNFPA, blank labelsNFPA, blank labelsPlastic—28,575×28,575 mm1000 labels/rollLog in for availability1 * 1 ROU137,00 €