Orders LinkContactUsLinkComponent
Face shield visor, 701.00
Face shield visor, 701.00
Interchangeable visor, Anti-fog, Acetate
Catalog # UNVT2200014
Supplier:   UNIVET
Face shield visor, 701.00
Catalog # UNVT2200014
Supplier:  UNIVET
Face shield visor, 701.00
Face shield visor, 701.00
Interchangeable visor, Anti-fog, Acetate
Price
70,50 €
Connexion Pour la disponibilité
1 * 5 ST
Product Details & Documents
The 701.00 face shield provides protection while ensuring comfort with a soft latex-free sponge on the forehead.
  • Anti-fog acetate visor
  • Soft latex-free sponge on the forehead
  • Adjustable elastic head band
  • Visor inclines up to 90°
  • Interchangeable visor
  • Face shield suitable for use in combination with prescription glasses and protective masks
The thermo-formed acetate screen maximizes vision due to anti-fog characteristics. The visor can be tilted up to an angle of 90 degrees, and is easily replaceable. The 701.00 visor is compatible with respiratory protective masks and corrective goggles.
Spécifications
  • Type:
    Anti-fog
  • Description:
    Interchangeable visor
  • Material:
    Acetate