ESD safety ankle boots, pull-on, S3, jo_Clean Slipper Mid, Jori
Supplier: ELTEN
Certificates
About this item
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
- Full bellows tongue
- MONO-PU sole jo_MONO
- Composite toe cap
- Metal- and leather-free
- Available in black or white finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certifications: EN ISO 20345 S3 SRC, Form B.
Specifications
- Catalog No:
- ELTN16671/48
- ELTN16671/42
- ELTN16671/43
- ELTN16671/44
- ELTN16671/45
- ELTN16671/46
- ELTN16671/47
- ELTN16661/36
- ELTN16661/37
- ELTN16661/38
- ELTN16661/39
- ELTN16661/40
- ELTN16661/41
- ELTN16671/38
- ELTN16671/39
- ELTN16671/40
- ELTN16671/41
- ELTN16661/42
- ELTN16661/43
- ELTN16671/36
- ELTN16661/44
- ELTN16671/37
- ELTN16661/45
- ELTN16661/46
- ELTN16661/47
- ELTN16661/48
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001