Nitrogen generator, Infinity 1051
Infinity 1051 nitrogen generator
Catalog # 564-0283
Supplier: PEAK SCIENTIFIC
Jump to:
Product Details & Documents
Designed and engineered as an on-site, compressor less nitrogen generation solution tailored for the Shimadzu LCMS-8045, 8050 or 8060. The Infinity 1051 delivers one output of nitrogen and another of dry air to laboratories that already have an existing air supply.
- Both a nitrogen and dry air output suitable for the Shimadzu LCMS-8045/50/60 and instruments requiring similar specifications
- 24/7 operation at optimum performance if required
- Gas is supplied on demand so generator works to your schedule
- Few moving parts means little maintenance required and ensures long life of the generator
- Minimal set-up required
- Completely silent in operation
- 1 year comprehensive on-site warranty
Specifications
- Description:Infinity 1051 nitrogen generator
- Cat. No.:564-0283