ContactUsLinkComponent
Grade 520 bII ½ filter papers for technical use, Whatman™
Grade 520 bII ½ filter papers for technical use, Whatman™
Certificates
undefined
Grade 520 bII ½ filter papers for technical use, Whatman™
Specifications
  • Catalog No:
  • 515-8036
  • 515-8038
  • 515-8030
  • WHAT10331553
  • 515-8064
  • Grade:
  • 520 bII ½
  • 520 bII ½
  • 520 bII ½
  • 520 bII ½
  • 520 bII ½
  • Weight:
  • 155 g/m²
  • 155 g/m²
  • 155 g/m²
  • 155 g/m²
  • 155 g/m²
  • Thickness:
  • 550 µm
  • 550 µm
  • 550 µm
  • 550 µm
  • 550 µm
  • Particle retention:
  • 20 µm
  • 20 µm
  • 20 µm
  • 20 µm
  • 20 µm
  • Filtration speed:
  • 30 (Herzberg)
  • 30 (Herzberg)
  • 30 (Herzberg)
  • 30 (Herzberg)
  • 30 (Herzberg)