Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Grade shark skin filter papers for technical use, Whatman™

Grade shark skin filter papers for technical use, Whatman™
Certificates
Grade shark skin filter papers for technical use, Whatman™
Grade shark skin filter papers for technical use, Whatman™

14 Options Available Below

Product Details & Documents

Whatman Grade Shark Skin filter papers are a creped filter paper that is commonly used to remove impurities in a variety of applications.

  • Very thin with high wet strength: Enables handling when wet; allows use under pressure; provides durability and structural support in aqueous filtration
  • Creped filter paper: Has enlarged surface area that reduces duration of filtration and allows more stretch and flexibility
  • Durable: Resistant to weak acids and bases

Whatman Grade Shark Skin Filter Papers (particle retention 8-12 µm) are a medium/slow filter paper made from super-refined cellulose. They are available in a variety of sizes, in both circles and sheets.

Applications include:
- Purification of edible oils, beverages, and cocoa butter for food and cosmetic uses.
- Removal of suspended solids for purification of electroplating baths
Protective paper for filter press cloths
- Reducing impurities lowers the amounts of reactants and increases the efficiency of the oil, which minimizes production costs and helps to ensure regulatory compliance.

 

Specifications for Product Family

Specifications

  • Catalog No:
  • WHAT10347509
  • WHAT10347510
  • WHAT10347511
  • WHAT10347513
  • WHAT10347512
  • WHAT10347519
  • WHAT10347521
  • WHAT10347577
  • WHAT10538877
  • WHAT10347530
  • WHAT10347522
  • WHAT10347523
  • WHAT10347525
  • WHAT10537138
  • Grade:
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Shark skin
  • Weight:
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • 44 g/m²
  • Thickness:
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • Particle retention:
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • 8 - 12 µm
  • Filtration speed:
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)
  • 77,5 s (Herzberg)

Specifications

Product Family Options

Product Information

  • Grade
    Size
    Availability
    Price
  • Shark skin
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 53,50 €
    53,50 €
    1 * 100 ST
       53,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347509
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 56,30 €
    56,30 €
    1 * 100 ST
       56,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347510
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 60,50 €
    60,50 €
    1 * 100 ST
       60,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347511
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,40 €
    53,40 €
    1 * 1 ST
       53,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347513
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 65,90 €
    65,90 €
    1 * 100 ST
       65,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347512
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 71,30 €
    71,30 €
    1 * 100 ST
       71,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347519
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    270 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 81,50 €
    81,50 €
    1 * 100 ST
       81,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347521
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    290 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 82,40 €
    82,40 €
    1 * 100 ST
       82,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347577
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    300×250 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 99,10 €
    99,10 €
    1 * 100 ST
       99,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10538877
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    320 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 85,20 €
    85,20 €
    1 * 100 ST
       85,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347530
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    340 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 87,40 €
    87,40 €
    1 * 100 ST
       87,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347522
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    385 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 93,50 €
    93,50 €
    1 * 100 ST
       93,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347523
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    500 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 156,00 €
    156,00 €
    1 * 100 ST
       156,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10347525
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)
  • Shark skin
    530 mm × 228,6 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 796,00 €
    796,00 €
    1 * 1 ST
       796,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10537138
    Grade
    Shark skin
    Weight
    44 g/m²
    Thickness
    170 µm
    Particle retention
    8 - 12 µm
    Filtration speed
    77,5 s (Herzberg)