Continuous level control, SymLine®
Supplier: S.C.A.T.
Jump to:
About this item
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
- Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
- Control box provides 5 freely configurable outlets to control external devices
- Disc sensor for level control from non-conductive material is easy to install and adjustable in height
- Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
- Canister made from electrostatic conductive HDPE material
The compact design and independent power supply allow simple handling and installation, also outside switch cabinets.
6 Options Available Below
Product Details & Documents
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
- Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
- Control box provides 5 freely configurable outlets to control external devices
- Disc sensor for level control from non-conductive material is easy to install and adjustable in height
- Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
- Canister made from electrostatic conductive HDPE material
The compact design and independent power supply allow simple handling and installation, also outside switch cabinets.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
TDS Canisters Overview
Datasheet
Product Family Options
Product Information
- TypeAvailabilityPrice
- SCTV108042-S1Safety canister, S60/61, with holder for sensor (SCTV108178)Safety canister, S60/61, with holder for sensor (SCTV108178)With holder for sensor (SCTV108178)Log in for availability1 * 1 ST172,00 €
- SCTV108178Capacitive rod sensor for the level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMURCapacitive rod sensor for the level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMURLog in for availability1 * 1 ST284,00 €
- SCTV160123Floater, electronic, 139 mm
- SCTV199005Protective basket for mechanical level control, suitable for all canisters with floaterProtective basket for mechanical level control, suitable for all canisters with floaterLog in for availability1 * 1 ST32,20 €
- 216-0923Protective basket for floater, PP, suitable for all electro conductive canisters with floaterProtective basket for floater, PP, suitable for all electro conductive canisters with floaterLog in for availability1 * 1 ST37,60 €
- SCTV502042Set for continuous level control with Signalbox UM-O-Ex with 230 VACSet for continuous level control with Signalbox UM-O-Ex with 230 VAC—Log in for availability1 * 1 ST1 650,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Built-in signal box table stand, brushed stainless steel, 2 mm wall thicknessBuilt-in signal box table stand, brushed stainless steel, 2 mm wall thicknessSCTV106698Log in for availability1 * 1 ST - 61,80 €
Product Information
- Catalog #AvailabilityPrice
- Built-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plugBuilt-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plugSCTV106548Log in for availability1 * 1 ST - 485,00 €
Product Information
- Catalog #AvailabilityPrice
- Built-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supplyBuilt-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supplySCTV106741Log in for availability1 * 1 ST - 323,00 €
Product Information
- Catalog #AvailabilityPrice
- Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)Built-in signal box display, brushed stainless steel, 2 mm wall thickness, for signal boxes 1/2/5 with cable connection on the back (Version 2024)SCTV106733Log in for availability1 * 1 ST - 61,80 €
Product Information
- Catalog #AvailabilityPrice
- Built-in Signalbox5, EU/UK/US, 5-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for five containers, five switch inputs and five switch outputs for peripheral units, including 24 V power supplyBuilt-in Signalbox5, EU/UK/US, 5-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for five containers, five switch inputs and five switch outputs for peripheral units, including 24 V power supplySCTV106735Log in for availability1 * 1 ST - 614,00 €



