ContactUsLinkComponent
Trace elements C, Corning®
Trace elements C, Corning®
Catalog # MDTC25-023-CI
Supplier:  Corning
CAS Number:  
undefined
Trace elements C, Corning®
Catalog # MDTC25-023-CI
Supplier:  Corning
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Trace elements C
Specifications
About this item
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.