ContactUsLinkComponent
Trace elements B, Corning®
Trace elements B, Corning®
Catalog # MDTC25-022-CI
Supplier:  Corning
CAS Number:  
undefined
Trace elements B, Corning®
Catalog # MDTC25-022-CI
Supplier:  Corning
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Trace elements B
Specifications
About this item
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.