Order Entry
Belgium
ContactUsLinkComponent
Chromocult® RAMBACH™ Agar ISO 6579 for Selective Isolation of Salmonella spp., Millipore
Chromocult® RAMBACH™ Agar ISO 6579 for Selective Isolation of Salmonella spp., Millipore
Supplier:  Merck
Certificates
Chromocult® RAMBACH™ Agar ISO 6579 for Selective Isolation of Salmonella spp., Millipore
Supplier:  Merck
Certificates

Specifications

  • Form:
    Powder
  • Key applications:
    Selective for enterobacteriaceae: Salmonella spp.
  • M-Clarity quality level:
    MQ200
  • Storage temperature:
    15-25°C
  • Application:
    Microbiological culture
  • pH:
    7.3±0.2 (25 °C)

Specifications

Related Information

Frequently Bought Together