Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-purpose scissors, Rubis®

Multi-purpose scissors, Rubis®

Straight, blunt/blunt, 147 mm, 0,5 mm
Catalog # 233-0218
Supplier:   OUTILS RUBIS
Multi-purpose scissors, Rubis®
Catalog # 233-0218
Supplier:  OUTILS RUBIS
Multi-purpose scissors, Rubis®
Multi-purpose scissors, Rubis®

Straight, blunt/blunt, 147 mm, 0,5 mm

Price
47,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Long legged multi-purpose scissors with rounded-off tips.

  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • Ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product

Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under a magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.

Specifications
  • Type:
    Straight, blunt/blunt
  • Length:
    147 mm
  • Tip size:
    0,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.