ContactUsLinkComponent
Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Supplier:  Halyard
Certificates
undefined
Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Supplier:  Halyard
Certificates
Specifications
  • Catalog No:
  • 112-6496
  • 112-6497
  • 112-6494
  • 112-6495
  • 112-6500
  • 112-6501
  • 112-6498
  • 112-6499
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Textured fingertips and palms
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • Palm thickness:
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +