Order Entry
Belgium
ContactUsLinkComponent
HyClone™ Fetal Bovine Serum, Characterized (New Zealand), HyClone Products (Cytiva)
HyClone™ Fetal Bovine Serum, Characterized (New Zealand), HyClone Products (Cytiva)
Certificates
undefined
HyClone™ Fetal Bovine Serum, Characterized (New Zealand), HyClone Products (Cytiva)

Specifications

  • Catalog No:
  • SH30406.02IR30
  • HYCLSH30406.02
  • SH30406.02IR35
  • SH30406.02IR25
  • SH30406.02IH25
  • SH30406.02-45
  • HYCLSH30406.02HI
  • Origin:
  • NZ
  • NZ
  • NZ
  • NZ
  • NZ
  • NZ
  • NZ
  • Endotoxin:
  • ≤20 EU/ml
  • ≤20 EU/ml
  • ≤20 EU/ml
  • ≤20 EU/ml
  • ≤20 EU/ml
  • ≤20 EU/ml
  • ≤20 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

Specifications

Frequently Bought Together