ContactUsLinkComponent
Wire weight, OIML F1
Wire weight, OIML F1
Catalog # SARBYCW1131-00
Supplier:  Sartorius Balances
undefined
Wire weight, OIML F1
Catalog # SARBYCW1131-00
Supplier:  Sartorius Balances
Specifications
  • Weight:
    10 mg
  • Description:
    Wire weight, OIML F1
  • Pk:
    1
  • No. of weights:
    1
Specifications
About this item
Accurate metrological weight with nominal mass value of 10 mg.
  • OIML class F1
  • Hardness: ca. 110 HB
  • Density of 7,950 kg/m³
  • High gloss polished surface
  • No laser markings
  • Without certificate
Certifications: Weights comply with the International Recommendation OIML-R111.