Orders LinkContactUsLinkComponent
Biemer vessel spring scissors
Biemer vessel spring scissors
Marketsource
Supplier:   FINE SCIENCE TOOLS
Biemer vessel spring scissors
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Biemer vessel spring scissors
Biemer vessel spring scissors
2 Options Available Below
Product Details & Documents
These stainless steel scissors are designed to securely cut vessels without slipping.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Availability
    Price
  • Blunt, straight tips
    85 mm
    4 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 238,50 €
    238,50 €
    1 * 1 Pce
       238,50 €
    Specifications:
    Marketsource
    Marketsource
  • Blunt, straight tips
    130 mm
    2 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 286,00 €
    286,00 €
    1 * 1 Pce
       286,00 €
    Specifications:
    Marketsource
    Marketsource