Orders LinkContactUsLinkComponent
Micro scissors
Micro scissors
Marketsource
Supplier:   FINE SCIENCE TOOLS
Micro scissors
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Micro scissors
2 Options Available Below
Product Details & Documents
These scissors are made of stainless steel.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Serrated
    Availability
    Price
  • Sharp, angled to side tips
    145 mm
    2,5 mm
    0,175 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 410,50 €
    410,50 €
    1 * 1 Pce
       410,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    145 mm
    2,5 mm
    0,175 mm
    +
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 413,50 €
    413,50 €
    1 * 1 Pce
       413,50 €
    Specifications:
    Marketsource
    Marketsource