Orders LinkContactUsLinkComponent
Moria spring scissors
Moria spring scissors
Marketsource
Supplier:   FINE SCIENCE TOOLS
Moria spring scissors
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Moria spring scissors
Moria spring scissors
2 Options Available Below
Product Details & Documents
These scissors are made of inox and have a very fine tip diameter.
Hand fashioned using 40x magnification, these tips are especially useful when incising vessel walls for catheter insertion.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Availability
    Price
  • Sharp, curved tips
    80 mm
    5 mm
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 531,50 €
    531,50 €
    1 * 1 Pce
       531,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    80 mm
    5 mm
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 501,50 €
    501,50 €
    1 * 1 Pce
       501,50 €
    Specifications:
    Marketsource
    Marketsource