Orders LinkContactUsLinkComponent
Vannas spring scissors, round handled
Vannas spring scissors, round handled
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vannas spring scissors, round handled
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Vannas spring scissors, round handled
Vannas spring scissors, round handled
2 Options Available Below
Product Details & Documents
These stainless steel scissors have round handles that allow easy fingertip adjustments.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Availability
    Price
  • Sharp, curved tips
    125 mm
    6 mm
    0,075 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 330,50 €
    330,50 €
    1 * 1 Pce
       330,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    125 mm
    6 mm
    0,075 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 296,00 €
    296,00 €
    1 * 1 Pce
       296,00 €
    Specifications:
    Marketsource
    Marketsource