Orders LinkContactUsLinkComponent
Vitrectomy scissors
Vitrectomy scissors
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vitrectomy scissors
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Vitrectomy scissors
Vitrectomy scissors
2 Options Available Below
Product Details & Documents
For the finest microdissection, these stainless steel spring scissors have the smallest blades.
  • Autoclavable
The black handles are made of ribbed plastic to ensure a solid grip.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Tip size
    Availability
    Price
  • Sharp, angled to side tips
    140 mm
    1 mm
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 667,50 €
    667,50 €
    1 * 1 Pce
       667,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    140 mm
    2,2 mm
    0,15 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 677,50 €
    677,50 €
    1 * 1 Pce
       677,50 €
    Specifications:
    Marketsource
    Marketsource